Learn More
Cayman Chemical HydroxocobalamIn 5mg
SDP
Supplier:  Cayman Chemical 240995 MG
Encompass_Preferred
A biologically active form of Vitamin B12; increases the number of red blood cells by 2.5-fold over control in a mouse model of anemia when administered at a dose of 2 UG per day for 4 days; efficacious in animal models of cyanide poisoning
Catalog No. 50-293-3081