Learn More
Cayman Chemical hydroxy BupropIon 10mg
SDP
Supplier:  Cayman Chemical 900052210 MG
Encompass_Preferred
An analytical reference standard categorized as an antidepressant; a metabolite of bupropion; substitutes for D-amphetamine in rats in a two-lever drug discrimination test; intended for research and forensic applications
Catalog No. 50-289-4776