Learn More
Cayman Chemical Hydroxy DImetrIdazole 5mg
SDP
Supplier:  Cayman Chemical 162085 MG
Encompass_Preferred
A metabolite of dimetridazole, the nitroimidazole-based antibacterial and anticoccidial agent suspected to be carcinogenic and mutagenic to humans
Catalog No. 50-297-0747