Learn More
Medchemexpress LLC Hydroxy iloperidone | 133454-55-4 | MFCD19705003 | 100.0% | 428.50 | C24H29FN2O4 | 10 MG

Supplier: Medchemexpress LLC HYG000310MG
Hydroxy iloperidone is the principal hydroxylated metabolite of the atypical antipsychotic iloperidone, provided as a high-purity research compound for analytical and preclinical laboratory use.
- High purity suitable for analytical studies.
- Molecular formula C24H29FN2O4 and molecular weight 428.50.
- Available in small milligram packages for laboratory workflows.
- CAS number 133454-55-4 for unambiguous identification.
- Intended for research use only; not for human or veterinary use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.