Learn More
Medchemexpress LLC Hydroxy Itraconazole | 112559-91-8 | >99.8% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY12772S1MG
Encompass_Preferred
Hydroxy Itraconazole is an active metabolite of Itraconazole (ITZ), which functions as a triazole antifungal agent. This compound is intended for research use only.
  • Active metabolite of Itraconazole
  • Functions as a triazole antifungal agent
  • Intended for research use
Catalog No. 50-003-40178