Learn More
Medchemexpress LLC Hydroxy itraconazole | 112559-91-8 | 99.8% | 10 MG

Supplier: Medchemexpress LLC HY1277210MG
Hydroxy Itraconazole is an active metabolite of Itraconazole (ITZ), a triazole antifungal agent. It has reported antifungal activity in vitro and inhibits CYP3A. This product is for research use only.
- Active metabolite of itraconazole
- Triazole antifungal agent
- Exhibits antifungal activity in vitro
- Inhibits CYP3A
- Molecular weight: 721.63
- Chemical formula: C35H38Cl2N8O5
- Appearance: White to off-white solid
- Soluble in DMSO (83.33 mg/mL)
- Insoluble in H2O (< 0.1 mg/mL)
- Store solid at 4°C, protected from light
- Store in solvent at -80°C for 6 months, or -20°C for 1 month (protected from light)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.