Learn More
Medchemexpress LLC Hydroxy Itraconazole | 112559-91-8 | 99.8% | 100 UG

Supplier: Medchemexpress LLC HY12772S100UG
Hydroxy Itraconazole is the analytical standard of Hydroxy Itraconazole, intended for research and analytical applications.
- CAS number: 112559-91-8
- Molecular weight: 721.63
- Molecular formula: C35H38Cl2N8O5
- Purity (LCMS): 99.80%
- Appearance: White to off-white solid
- Storage: 4°C, protect from light (solid); -80°C, 6 months; -20°C, 1 month (in solvent)
- Application: Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.