Learn More
Cayman Chemical Hydroxy Itraconazole 500ug
SDP
Supplier:  Cayman Chemical 22576500 UG
Encompass_Preferred
A metabolite of itraconazole; exhibits in vitro antifungal activity (MIC = 0.019, 0.078, 0.078 mg/ml against C. albicans, A. fumigatus, and C. neoformanss, respectively); plasma concentration is used as an indicator of prophylaxis for fungal infections
Catalog No. 50-298-6735