Learn More
Medchemexpress LLC Hydroxy-PEG1-acid 5g | 89211-34-7 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1166555G
Encompass_Preferred
Hydroxy-PEG1-acid is a single-unit polyethylene glycol (PEG1) linker with terminal hydroxyl and carboxylic acid groups. Supplied for research use as a non-cleavable ADC linker, it provides a short PEG spacer to improve solubility and spacing between an antibody and payload in antibody-drug conjugates.
  • Non-cleavable single-unit PEG linker for ADC synthesis.
  • Terminal hydroxyl and carboxylic acid functional groups for conjugation.
  • Short PEG spacer to improve solubility and reduce aggregation.
  • Typical purity around 95% and colorless to light yellow appearance.
  • Available in small research pack sizes, including 5 G.
Catalog No. 50-002-27141