Learn More
Medchemexpress LLC Hydroxy-PEG3-2-methylacrylate | 2351-42-0 | MFCD30723235 | 98.0% | 218.25 | C10H18O5 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW1909465G
Encompass_Preferred
Hydroxy-PEG3-2-methylacrylate is a PEG-based hydroxy-functional methacrylate monomer used as a building block or linker in polymer synthesis, bioconjugation, and related life-science research. It offers a short PEG spacer with a terminal hydroxy group and a polymerizable 2-methylacrylate moiety, with lot-specific purity reported on the Certificate of Analysis.
  • Provides a short PEG3 spacer for flexible linkage.
  • Contains a hydroxy functional group for further derivatization.
  • Includes a polymerizable 2-methylacrylate moiety for radical polymerization.
  • Supplied in laboratory-scale pack sizes suitable for research use.
  • Lot-specific purity and storage conditions are available in the Certificate of Analysis.
Catalog No. 50-002-82217