Learn More
Medchemexpress LLC HY-42488 100mg Medchemexpress, Hydroxy-PEG3-(CH2)2-Boc CAS:186020-66-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY42488100MG
Encompass_Preferred
Medchemexpress, HY-42488 100mg Hydroxy-PEG3-(CH2)2-Boc CAS:186020-66-6 Hydroxy-PEG2-(CH2)2-Boc is a uncleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Hydroxy-PEG2-(CH2)2-Boc is extracted from patent WO2004008101A2 (compound 196). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0055