Learn More
Medchemexpress LLC Hydroxy-PEG4-acid | 5G
SDP
Supplier:  Medchemexpress LLC HY1171045G
Encompass_Preferred
Hydroxy-PEG4-acid is a non-cleavable 4 unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs) Hydroxy-PEG4-acid is also a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-003-08338