Learn More
Medchemexpress LLC Hydroxy-PEG4-(CH2)2-Boc | 518044-32-1 | MFCD11041117 | 99.3% | 322.39 g/mol | C15H30O7 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW039178100G
Encompass_Preferred
Hydroxy-PEG4-(CH2)2-Boc is a PEG4-based linker with a terminal hydroxyl and a tert-butyl (Boc) protected group, supplied for use in synthetic and medicinal chemistry applications such as antibody-drug conjugate (ADC) and PROTAC assembly. It is a non-cleavable spacer with a molecular weight of 322.39 g/mol and a chemical formula of C15H30O7.
  • PEG4 spacer with terminal hydroxyl
  • Boc-protected tert-butyl ester functional group
  • Non-cleavable linker for ADC and PROTAC assembly
  • Approximately 99.3% purity suitable for medicinal chemistry
  • Available in laboratory-scale pack sizes, including 100 G
  • Recommended storage: -20°C for long-term stability; follow solvent-specific conditions
Catalog No. 50-002-03140