Learn More
Medchemexpress LLC Hydroxy-PP-Me | 833481-77-9 | 99.9% | 282.34 g/mol | C16H18N4O | 10 MG

Supplier: Medchemexpress LLC HY15669410MG
Hydroxy-PP-Me is a selective small-molecule inhibitor of carbonyl reductase 1 (CBR1) used in cancer research. It inhibits serum starvation-induced apoptosis and can enhance the cytotoxic effects of daunorubicin and arsenic trioxide. Supplied as a high-purity analytical reagent, it is commonly provided as a 10 mg unit.
- High purity: 99.92%.
- Molecular formula C16H18N4O; molecular weight 282.34 g/mol.
- Identified CAS number 833481-77-9.
- Reported CBR1 IC50 ≈ 759 nM.
- Solubility: soluble in DMSO; available as 10 mM solutions in DMSO.
- For research use only; not for human or diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.