Learn More
Medchemexpress LLC HY-Y0882 500mg Medchemexpress, Hydroxyamine (hydrochloride) CAS:5470-11-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY0882500MG
Encompass_Preferred
Medchemexpress, HY-Y0882 500mg Hydroxyamine (hydrochloride) CAS:5470-11-1 Hydroxyamine hydrochloride is a selective monoamine oxidase (MAO) inhibitor used for inhibiting of platelet aggregation. Hydroxyamine hydrochloride is an intermediate of organic synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3552
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.