Learn More
Cayman Chemical β-Hydroxybutyrate (Ketone Body) Colorimetric Assay Kit, 96 wells
SDP
Supplier:  Cayman Chemical 70019096WELLS
Encompass_Preferred
β-HB is a "ketone body" which is produced in the liver, mainly from the oxidation of fatty acids, and is exported to peripheral tissues for use as an energy source. Normally ketosis can indicate that lipid metabolism has been activated and the pathway of lipid degradation is intact. Pathological causes of ketosis include multiple organ failure, diabetes, childhood hypoglycemia, corticosteroid or growth hormone deficiency, intoxication with alcohol or salicylates and several inborn errors of metabolism. Cayman’s β-HB (Ketone Body) Assay provides a simple, reproducible, and sensitive tool for measuring β-HB levels in plasma, serum, or urine in a 96-well plate format with a colorimetric readout at 445-455 nm.
Catalog No. NC9044966