Learn More
Biomatik Corporation Β-Hydroxybutyryl-Hist1H3A (K9) Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11531
Encompass_Preferred
Β-Hydroxybutyryl-Hist1H3A (K9) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to HIST1H3A. The Β-Hydroxybutyryl-Hist1H3A (K9) Polyclonal Antibody has been validated for the following applications: ELISA, WB, ICC, ChIP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Peptide Sequence Around Site Of Β-Hydroxybutyryl-Lys (9) Derived From Human Histone H3.1. Applications: ELISA, WB, ICC, ChIP. Target Synonyms: HIST1H3A. Accession Number: P68431. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9241