Learn More
Cayman Chemical HydroxychloroquIn-d4sulfat 1mg
SDP
Supplier:  Cayman Chemical 223751 MG
Encompass_Preferred
An internal standard for the quantification of hydroxychloroquine by GC- or LC-MS
Catalog No. 50-298-7799