Learn More
Selleck Chemical LLC Hydroxychloroquine (HCQ) Sulfate S4430-200mg
SDP
Supplier:  Selleck Chemical LLC S4430200MG
Encompass_Preferred
Hydroxychloroquine (HCQ) Sulfate is an antimalarial agent used for the treatment of systemic lupus erythematosus rheumatoid arthritis and other autoimmune inflammatory and dermatologic conditions Also acts as an inhibitor of autophagy and toll-like receptor (TLR) 7/9
Catalog No. 50-311-0884