Learn More
Apexbio Technology LLC Hydroxychloroquine Sulfate(Synonyms: Plaquenil, HCQ sulfate, Hydroxychloroquine sulphate, Hydroxychloroquine sulfate salt, HCQS), 10mM (in 1mL H2O), CAS: 747-36-4.
SDP
Supplier:  Apexbio Technology LLC B48745.1
Encompass_Preferred
Hydroxychloroquine Sulfate (CAS 747-36-4) is a synthetic quinoline derivative widely employed in biomedical research due to its immunomodulatory activities As an inhibitor of autophagy and toll-like receptors 7 and 9 (TLR7/9) it disrupts intracellular signaling pathways activated by nucleic acid ligands thus modulating dysregulated immune responses At a concentration of 5 M hydroxychloroquine sulfate blocks ligand-induced TLR7/9 activation without altering cellular pH levels It is commonly used in research contexts for studying autoimmune pathologies such as systemic lupus erythematosus and rheumatoid arthritis
Catalog No. 50-000-60919