Learn More
Medchemexpress LLC HY-113239 1mg Medchemexpress, Hydroxycotinine CAS:34834-67-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1132391MG
Encompass_Preferred
Medchemexpress, HY-113239 1mg Hydroxycotinine CAS:34834-67-8 Hydroxycotinine is the main nicotine metabolite detected in smokers urine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9234