Learn More
Medchemexpress LLC Hydroxyethyl starch | 9005-27-0 | MFCD00131356 | 100.7% | 170-230 kDa | (C6H10O5)m(C2H5O)n | 1 KG
SDP
Supplier:  Medchemexpress LLC HYW1455191KG
Encompass_Preferred
Hydroxyethyl starch (MW170-230 kDa) is a high-molecular-weight hydroxyalkylated polysaccharide used as a biochemical reagent and colloidal agent in life-science research. Supplied as a white to off-white solid, it is employed in formulations, volume-expander studies, and laboratory assays where modified starch polymers are required.
  • High molecular weight polymer, 170-230 kDa.
  • White to off-white solid for convenient handling and storage.
  • Analytical assay reported on COA for lot control.
  • Very low heavy metal content (<0.001%).
  • Low sodium chloride content (0.012%).
  • Intrinsic viscosity of 17.4 to support formulation consistency.
  • Suitable for formulations, colloid research, and laboratory assays.
Catalog No. 50-002-82924