Learn More
Medchemexpress LLC Hydroxyethyl starch | 9005-27-0 | MFCD00131356 | 100.7% | 170-230 kDa | (C6H10O5)m(C2H5O)n | 100 G
SDP
Supplier:  Medchemexpress LLC HYW145519100G
Encompass_Preferred
Hydroxyethyl starch is a high-molecular-weight polysaccharide derivative supplied as a white to off-white powder for laboratory research use. It functions as a biochemical reagent and formulation component, offering controlled molecular weight and aqueous solubility for experimental and assay applications.
  • High molecular weight (170-230 kDa) suitable for viscosity and colloidal applications.
  • Supplied as a solid powder, white to off-white in appearance.
  • Soluble in water at approximately 100 mg/mL with ultrasonic assistance.
  • Stable when stored as powder at -20°C for up to 3 years; in solution store at -80°C for extended storage.
  • Provided for research use only; not for clinical or patient use.
Catalog No. 50-002-82923