Learn More
Medchemexpress LLC Hydroxyethyl starch | 9005-27-0 | MFCD00131356 | 100.7% | 170-230 kDa | (C6H10O5)m(C2H5O)n | 50 G
SDP
Supplier:  Medchemexpress LLC HYW14551950G
Encompass_Preferred
Hydroxyethyl starch (MW 170-230 kDa) is a hydroxyethylated polysaccharide supplied for life-science research. It is provided as a white to off-white solid intended for use as a biochemical reagent or biological material when a defined high-molecular-weight starch derivative is required.
  • Defined molecular weight range 170-230 kDa.
  • White to off-white solid form with high assay purity.
  • Low heavy metal content (<0.001%).
  • Reported pH of approximately 5.3 on the COA.
  • Intrinsic viscosity 17.4 (as reported on COA).
  • Available in multiple pack sizes including 50 g for laboratory use.
Catalog No. 50-002-83088