Learn More
Medchemexpress LLC Hydroxyethyl starch | 9005-27-0 | MFCD00131356 | 170-230 kDa | (C6H10O5)m•(C2H5O)n | 500 G

Supplier: Medchemexpress LLC HYW145519500G
Hydroxyethyl starch is a hydroxyalkylated polysaccharide supplied as a solid biochemical assay reagent for laboratory research. It is commonly used as a colloidal volume expander, matrix component, or viscosity modifier and is characterized by a molecular weight range of 170-230 kDa and CAS number 9005-27-0.
- Polysaccharide with hydroxyethyl substitution suitable for biochemical assays.
- Molecular weight range 170-230 kDa for colloidal behavior and viscosity control.
- Solid form for convenient storage and handling.
- CAS number 9005-27-0 for unambiguous identification.
- Available in multiple pack sizes to support small- to large-scale experiments.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.