Learn More
Medchemexpress LLC Hydroxyethylthio vitamin K3 | 59147-84-1 | 248.30 g/mol | C13H12O3S | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW024644
Encompass_Preferred
Hydroxyethylthio vitamin K3 is a research-grade small molecule reported to inhibit Cdc25 phosphatases and used in cell-cycle and tumor research. It is characterized by molecular formula C13H12O3S and molecular weight 248.30 g/mol, and is supplied in milligram-scale quantities for biochemical and cellular studies.
  • Cdc25 phosphatase inhibitor with reported IC50 of 4.56 μM (Cdc25A) and 23.63 μM (Cdc25B).
  • Small molecule with formula C13H12O3S and molecular weight 248.30 g/mol.
  • Suitable for biochemical and cellular research applications.
  • Provided in milligram-scale pack sizes with recommended storage at room temperature or cold solvent conditions.
  • Solid appearance; handle according to standard laboratory safety practices.
Catalog No. 50-003-03284