Learn More
Medchemexpress LLC HY-13911 5mg Medchemexpress, Hydroxyfasudil CAS:105628-72-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY139115MG
Encompass_Preferred
Medchemexpress, HY-13911 5mg Hydroxyfasudil CAS:105628-72-6 Hydroxyfasudil is a ROCK inhibitor, with IC 50 s of 0.73 and 0.72 μM for ROCK1 and ROCK2 , respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2693