Learn More
Selleck Chemical LLC Hydroxyfasudil HCl
SDP
Supplier:  Selleck Chemical LLC S8208100MG
Encompass_Preferred
Hydroxyfasudil HCl an active metabolite of fasudil hydrochloride is a specific Rho-kinase(ROCK) inhibitor with IC50 values of 0 73 M and 0 72 M for ROCK1 and ROCK2 respectively
Catalog No. 50-000-02011