Learn More
Medchemexpress LLC Hydroxyflutamide | 52806-53-8 | 99.77% | 292.21 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0132725MG
Encompass_Preferred
Hydroxyflutamide (HFT) is the active metabolite of Flutamide (HY-B0022) and is orally active. It acts as a potent androgen receptor antagonist with an IC50 of 700 nM. This compound has been shown to affect embryonic development and reproductive tract development in mice. Additionally, Hydroxyflutamide can enhance the efficacy of Bacillus Calmette-Guérin (BCG) in inhibiting the progression of bladder cancer. It is suitable for research related to tumors and reproductive diseases.
  • Active metabolite of Flutamide.
  • Exhibits oral activity.
  • Potent androgen receptor antagonist with an IC50 of 700 nM.
  • Can affect embryonic development and reproductive tract development in mice.
  • Enhances the efficacy of Bacillus Calmette-Guérin (BCG) to inhibit bladder cancer progression.
  • Useful in research related to tumors and reproductive diseases.
Catalog No. 50-004-54239