Learn More
Medchemexpress LLC Hydroxylamine, O-[(4-nitrophenyl)methyl]-, hydrochloride (1:1) | 2086-26-2 | 99.8% | 204.61 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0124691G
Encompass_Preferred
O-(4-Nitrobenzyl)hydroxylamine hydrochloride is a biochemical assay reagent utilized for the creation of N-(4-nitrobenzyloxy)-amino acid, which serves as substrates for an unambiguous N-hydroxypeptide synthesis.
  • Biochemical assay reagent.
  • Used for the preparation of N-(4-nitrobenzyloxy)-amino acid.
  • Substrates for unambiguous N-hydroxypeptide synthesis.
Catalog No. 50-004-53888