Learn More
Medchemexpress LLC Hydroxylamine, O,O'-(3,6,9,12-tetraoxatetradecane-1,14-diyl)bis- | 98627-72-6 | 96.0% | C10H24N2O6 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140410
Encompass_Preferred
Bis-aminooxy-PEG4 is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs consist of two distinct ligands joined by a linker, one targeting an E3 ubiquitin ligase and the other for a specific target protein. This mechanism leverages the intracellular ubiquitin-proteasome system to achieve selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52735