Learn More
Medchemexpress LLC α-hydroxymethyl atropine | 2515-36-8 | MFCD00067306 | >=98.0% | 319.40 | C18H25NO4 | 10 MG

Supplier: Medchemexpress LLC HYW58792710MG
α-Hydroxymethyl atropine is an impurity that can form during atropine synthesis, supplied as a research-grade analytical reference for impurity profiling, method development, and validation in laboratory settings.
- Intended for use as an analytical impurity reference.
- High purity ≥98.0% suitable for analytical applications.
- Molecular weight 319.40, formula C18H25NO4.
- CAS number 2515-36-8 for unambiguous identification.
- Available in small quantities suitable for laboratory testing.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.