Learn More
Medchemexpress LLC α-hydroxymethyl atropine | 2515-36-8 | MFCD01733106 | ≥98.0% | 319.40 | C18H25NO4 | 5 MG

Supplier: Medchemexpress LLC HYW5879275MG
α-Hydroxymethyl atropine is an impurity produced during atropine synthesis and is supplied as a high-purity analytical reference for laboratory research. It is intended for use in impurity profiling, method development, and synthesis characterization; not for human or veterinary use.
- High purity: ≥98.0%
- Cas number: 2515-36-8
- Molecular weight: 319.40
- Suitable as an analytical reference or impurity standard
- Useful for impurity profiling and method development
- Supplied as a 5 MG sample for laboratory use
- For research use only; not for human or veterinary use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.