Learn More
Cayman Chemical HydroxymetronIdazole 10mg
SDP
Supplier:  Cayman Chemical 3524910 MG
Encompass_Preferred
An active metabolite of metronidazole; active against B. fragilis, B. thetaiotaomicron, B. distasonis, and B. ovatus (MICs = 1, 2, 1, and 2 UG/ml, respectively); has been found in pork and porcine plasma
Catalog No. 50-291-3690