Learn More
Medchemexpress LLC Hydroxyphenyl Fluorescein (HPF) | 359010-69-8 | 99.9% | 424.40 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY11133010MG
Encompass_Preferred
Hydroxyphenyl Fluorescein (HPF) is a stable Reactive Oxygen Species (ROS) fluorescent probe dye. It exhibits stronger specificity and stability compared to H2DCFDA. Hydroxyphenyl Fluorescein generates strong green fluorescence through a hydroxyl radical reaction with intracellular peroxynitroso. It can be utilized for fluorescence microscopy, high-throughput imagers, luciferase microplate readers, or flow cytometry. The excitation/emission wavelengths are 490/515 nm.
  • Stable ROS fluorescent probe dye.
  • Stronger specificity and stability than H2DCFDA.
  • Produces strong green fluorescence via hydroxyl radical reaction with intracellular peroxynitroso.
  • Applicable for fluorescence microscopy, high-throughput imager, luciferase microplate reader, or flow cytometry.
  • Ex/Em=490/515 nm.
Catalog No. 50-003-46463