Learn More
Medchemexpress LLC Hydroxyprogesterone caproate (Standard) | 630-56-8 | MFCD00072134 | 99.2% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB0742R
Encompass_Preferred
Hydroxyprogesterone caproate (Standard) is the analytical standard of Hydroxyprogesterone caproate. This product is intended for research and analytical applications. It is supplied as a solid with a white to off-white color and is for research use only.
  • Intended for research and analytical applications
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS
  • Reference standard supplied assay
Catalog No. 50-004-06517