Learn More
Medchemexpress LLC Hydroxypropyl cellulose | 9004-64-2 | 97.0% | C6H7O2(OR1)(OR2)(OR3) | 100 G
SDP
Supplier:  Medchemexpress LLC HY100557100G
Encompass_Preferred
Hydroxypropyl cellulose is a cellulose ether excipient used as a formulation aid and viscosity modifier in pharmaceutical and laboratory applications. It is a white to off-white powder that provides water solubility, film-forming properties, and thickening performance in aqueous and non-aqueous systems. Typical uses include coatings, suspensions, emulsions, and tablet binding. Typical purity is about 97.0% (NMR). CAS 9004-64-2.
Catalog No. 50-002-13463