Learn More
TARGETMOL CHEMICALS INC HYDROXYSTILBAMIDINE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T98795MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Hydroxystilbamidine ((E)-hydroxystilbamidine) a dye capable of binding to both DNA and RNA has been found to be a powerful inhibitor of cellular ribonucleases. purity: 98%
Catalog No. 50-257-8943