Learn More
Aat Bioquest Hydroxystilbamidine CAS 223769-64-0 (10 mg)
SDP
Supplier:  Aat Bioquest 17514
Encompass_Preferred
Hydroxystilbamidine also called Fluoro Gold is used for staining DNA and RNA. This catalog is for 10 mg.
Catalog No. 50-224-9720