Learn More
Medchemexpress LLC HY-108166A 1mg Medchemexpress, Hydroxystilbamidine bis(methanesulfonate) CAS:223769-64-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY108166A1MG
Encompass_Preferred
Medchemexpress, HY-108166A 1mg Hydroxystilbamidine bis(methanesulfonate) CAS:223769-64-0 Hydroxystilbamidine bis(methanesulfonate), a dye capable of binding to both DNA and RNA, has been found to be a powerful inhibitor of cellular ribonucleases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0730