Learn More
Medchemexpress LLC Hydroxytyrosol acetate | 69039-02-7 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN604310MM/1ML
Encompass_Preferred
Hydroxytyrosol acetate is an orally active, blood-brain barrier-permeable multi-active compound with diverse effects, including antibacterial, antioxidant, anti-platelet aggregation, and neuroprotective activities. It functions as a prodrug for Hydroxytyrosol, being converted to its active form in vivo. This compound is extensively used in scientific research concerning *Vibrio* infection, arterial thrombosis, Alzheimer's disease, and other related fields.
  • Inhibits the growth of *Vibrio* bacteria by increasing membrane permeability and interacting with DNA.
  • Reduces thrombosis and inhibits lipid oxidation by inhibiting COX activity and promoting vascular nitric oxide production.
  • Alleviates neuronal apoptosis and inflammatory responses by up-regulating ERβ expression.
  • Improves cognitive function in Alzheimer's disease models.
  • Shows time-dependent DPPH radical scavenging activity.
Catalog No. 50-004-18879