Learn More
Cayman Chemical HygromycIn A 25mg
SDP
Supplier:  Cayman Chemical 3556525 MG
Encompass_Preferred
An antibiotic; active against numerous spirochete bacteria, including T. pallidum, L. biflexa, and four strains of B. burgdorferi (MICs = 0.03-4 UG/ml for all); binds to the PTC in the 70S ribosomal subunit to inhibit protein synthesis; completely eliminates bacterial infection in skin biopsies from B. burgdorferi-infected mice in a model of acute Lyme disease from 50-250 mg/kg twice per day
Catalog No. 50-290-9694