Learn More
Medchemexpress LLC HYNIC-iPSMA TFA | 2375849-11-7 | 98.1% | 765.69 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP5292A10MG
Encompass_Preferred
HYNIC-iPSMA TFA is a ligand designed for the molecular imaging of tumors. This compound is comprised of two key components: HYNIC (6-hydrazinonicotinamide) and iPSMA (Inhibitor of Prostate-Specific Membrane Antigen). HYNIC serves as a chemical linker, utilized for attaching radioactive isotopes to specific targeted molecules. iPSMA acts as a selective inhibitor, designed to target and inhibit prostate-specific membrane antigen (PSMA).
  • Tumor molecular imaging.
  • Component-based design.
  • Radioactive isotope attachment.
  • Prostate-specific membrane antigen inhibition.
  • Prostate cancer detection.
  • High specificity and sensitivity.
  • Radionuclide-drug conjugate synthesis.
Catalog No. 50-003-98015