Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HyNic-PEG2-DBCO | 2741281-12-7 | >98.0% | 610.72 g/mol | C34H38N6O5 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1334985MG

Encompass_Preferred

HyNic-PEG2-DBCO is a bifunctional PEG2 linker combining a hydrazinonicotinamide (HyNic) reactive handle with a dibenzocyclooctyne (DBCO) moiety for orthogonal bioconjugation. Supplied as a solid (5 mg), it is intended for research use in PROTAC synthesis, protein/antibody conjugation, and copper-free click-chemistry workflows.

  • Provides orthogonal conjugation: hydrazone formation and copper-free SPAAC.
  • Contains HyNic and DBCO reactive groups for selective coupling.
  • PEG2 spacer improves solubility and reduces steric hindrance.
  • High purity (>98%) suitable for research applications.
  • Supplied as a solid powder for straightforward storage and handling.
  • Recommended storage refrigerated at 2-8 °C to preserve stability.

Catalog No. 50-002-62685


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.