Learn More
Medchemexpress LLC Hyocholic acid | 547-75-1 | 99.5% | 5 MG

Supplier: Medchemexpress LLC HY1212385MG
Hyocholic acid is a trihydroxy-5β-cholanic primary bile acid used as a research reagent in biochemical and metabolic studies. It contains hydroxy groups at the 3α-, 6α-, and 7α-positions and is provided for analytical assays and pathway investigations.
- Purity 99.46% as reported by supplier.
- Molecular weight 408.6 g/mol.
- Chemical formula C24H40O5.
- Available as a 5 mg solid and as 10 mM solutions in DMSO.
- Suitable for metabolic, bile acid, and signaling pathway studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.