Learn More
Medchemexpress LLC Hyocholic acid | 547-75-1 | 99.5% | 50 MG

Supplier: Medchemexpress LLC HY12123850MG
Hyocholic acid (CAS 547-75-1) is a trihydroxy bile acid used in biochemical and bile-acid pathway research. Supplied as a white to light-yellow solid with high analytical purity, it is suitable for in vitro studies and biochemical assays when handled and stored under recommended conditions.
- Purity 99.46%.
- Chemical formula C24H40O5; molecular weight 408.6 g/mol.
- Supplied as a 50 MG vial and in solution forms (10 mM in DMSO).
- Appearance: solid, white to light yellow.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.