Learn More
Cayman Chemical HyocholIc AcId-d4 5mg
SDP
Supplier:  Cayman Chemical 270335 MG
Encompass_Preferred
An internal standard for the quantification of hyocholic acid by GC- or LC-MS
Catalog No. 50-294-1531