Learn More
Cayman Chemical HyodeoxycholIc AcId 50g
SDP
Supplier:  Cayman Chemical 2029450 G
Encompass_Preferred
A secondary bile acid; decreases plasma VLDL and LDL cholesterol levels and reduces fasting glucose levels and atherosclerotic lesion size in LDL receptor knockout mice fed a Western diet at 1.25% in the diet; serum levels are increased in patients with Crohn’s disease or ulcerative colitis
Catalog No. 50-298-4707