Learn More
Medchemexpress LLC HYOU1 Human HEK293 500ug
SDP
Supplier:  Medchemexpress LLC HYP70943500UG
Encompass_Preferred
Recombinant human HYOU1 (hypoxia up-regulated protein 1) is supplied as a His-tagged protein expressed in HEK293 cells and provided as a lyophilized powder for research use. The preparation is characterized for endotoxin level and ATPase activity, and is formulated in PBS for reliable reconstitution and long-term storage.
  • Recombinant human protein expressed in HEK293 cells for native-like glycosylation.
  • His-tagged for convenient detection and purification applications.
  • Lyophilized powder formulation for extended shelf life and easy shipping.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Characterized ATPase activity with reported specific activity for functional studies.
  • Provides recommendations for reconstitution and storage to preserve activity.
Catalog No. 50-001-96859