Learn More
TARGETMOL CHEMICALS INC HYPACONITINE 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6S063010MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Hypaconitine an active and highly toxic constituent derived from Aconitum species has anti-inflammatory activity is widely used to treat rheumatism. purity: 99%
Catalog No. 50-257-6387